Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-O-Isopropylidene-D-glucal, 97%, Thermo Scientific Chemicals

Catalog No. AAH6022103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6022103 1 g
AAH60221MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6022103 Supplier Thermo Scientific Chemicals Supplier No. H6022103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

4,6-O-Isopropylidene-D-glucal is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-O-Isopropylidene-D-glucal is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51450-36-3
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
MDL Number MFCD22988998
InChI Key GAQDIYMHBQNXLE-PRJMDXOYSA-N
Synonym 4,6-o-isopropylidene-d-glucal, 4-o,6-o-isopropylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,2-didehydro-1,2-dideoxy-4-o,6-o-isopropylidene-beta-d-glucopyranose, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol
PubChem CID 11052343
IUPAC Name (4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
SMILES CC1(OCC2C(O1)C(C=CO2)O)C

Specifications

Quantity 1 g
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4,6-O-Isopropylidene-D-glucal
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.