Learn More
Medchemexpress LLC 4,7,10,13-tetraoxahexadecanedioic acid | 31127-85-2 | 98.0% | 500 MG

Supplier: Medchemexpress LLC HY119429500MG
Bis-PEG4-acid is a short polyethylene glycol (PEG) linker bearing two terminal carboxylic acid groups, designed for use in PROTAC synthesis and bioconjugation workflows.
- Short PEG spacer with two terminal carboxylic acids.
- Purity ≥98.0% by HPLC.
- Molecular weight 294.30 g/mol and formula C12H22O8.
- Colorless to off-white solid-liquid mixture.
- Improves solubility and provides convenient coupling points for sequential conjugation.
- Stable when stored at -20°C; in solvent store at -80°C for extended preservation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.