Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7-Di(2-thienyl)-2,1,3-benzothiadiazole 98.0+%, TCI America™
SDP

Catalog No. D44871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44871G 1 g
D4487200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44871G Supplier TCI America Supplier No. D44871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 165190-76-1
Molecular Formula C14H8N2S3
Molecular Weight (g/mol) 300.412
MDL Number MFCD18252231
InChI Key XGERJWSXTKVPSV-UHFFFAOYSA-N
PubChem CID 396294
IUPAC Name 4,7-dithiophen-2-yl-2,1,3-benzothiadiazole
SMILES C1=CSC(=C1)C2=CC=C(C3=NSN=C23)C4=CC=CS4

Specifications

Melting Point 124°C
Quantity 1 g
Formula Weight 300.41
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Di(2-thienyl)-2,1,3-benzothiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.