Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine 97.0+%, TCI America™
SDP

Catalog No. D44421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44421G 1 g
D4442200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44421G Supplier TCI America Supplier No. D44421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 333432-27-2
Molecular Formula C5HBr2N3S
Molecular Weight (g/mol) 294.952
MDL Number MFCD23703147
InChI Key LEHZIBSAFRVAJP-UHFFFAOYSA-N
PubChem CID 58129789
IUPAC Name 4,7-dibromo-[1,2,5]thiadiazolo[3,4-c]pyridine
SMILES C1=C(C2=NSN=C2C(=N1)Br)Br

Specifications

Melting Point 148°C
Color Yellow
Quantity 1 g
Formula Weight 294.95
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.