Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7-Dibromo-3-hydroxy-2-naphthoic Acid 98.0+%, TCI America™
SDP

Catalog No. D36465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36465G Supplier TCI America Supplier No. D36465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1779-10-8
Molecular Formula C11H6Br2O3
Molecular Weight (g/mol) 345.97
MDL Number MFCD00046367
InChI Key WNMKUIQCIRAXBN-UHFFFAOYSA-N
Synonym 4,7-dibromo-3-hydroxy-2-naphthoic acid, 1,6-dibromo-2-hydroxynaphthalene-3-carboxylic acid, 2-naphthalenecarboxylic acid, 4,7-dibromo-3-hydroxy, 1,6-dibromo-2-naphthol-3-carboxylic acid, 2-naphthoic acid,7-dibromo-3-hydroxy, 2-naphthalenecarboxylic acid,7-dibromo-3-hydroxy, acmc-1btqt, 2-naphthoic acid, 4,7-dibromo-3-hydroxy, labotest-bb lt00455402, 1,6-dibromo-2-hydroxynaphthalene-3-carb
PubChem CID 74502
IUPAC Name 4,7-dibromo-3-hydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=C(O)C(Br)=C2C=CC(Br)=CC2=C1

Specifications

Melting Point 253°C
Quantity 5 g
Formula Weight 345.97
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo-3-hydroxy-2-naphthoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.