Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamido-1-benzylpiperidine 98.0+%, TCI America™
SDP

Catalog No. A165425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A165425G 25 g
A16545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A165425G Supplier TCI America Supplier No. A165425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50534-23-1
Molecular Formula C14H20N2O
Molecular Weight (g/mol) 232.33
MDL Number MFCD00247343
InChI Key PXKZVCOKJFOTQY-UHFFFAOYSA-N
PubChem CID 12641850
IUPAC Name N-(1-benzylpiperidin-4-yl)acetamide
SMILES CC(=O)NC1CCN(CC2=CC=CC=C2)CC1

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 25 g
Formula Weight 232.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamido-1-benzylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.