Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidobutyric acid, 99%, Thermo Scientific™

Catalog No. AAA1103930
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1103930 250 g
AAA1103909 10 g
AAA1103918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1103930 Supplier Thermo Scientific Chemicals Supplier No. A1103930

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Acetamidobutyric acid is used as pharmaceutical intermediate.

Solubility
Soluble in water (19.9 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3025-96-5
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00020552
InChI Key UZTFMUBKZQVKLK-UHFFFAOYSA-N
Synonym 4-acetamidobutyric acid, 4-acetylamino butanoic acid, n-acetyl-4-aminobutyric acid, n-acetyl-4-aminobutanoic acid, n-acetyl-gamma-aminobutyrate, butyric acid, 4-acetamido, 4-acetamidobutanoate, 4-acetamidobutyrate, gamma-acetylaminobutyric acid, n4-acetylaminobutanoate
PubChem CID 18189
ChEBI CHEBI:17645
IUPAC Name 4-acetamidobutanoic acid
SMILES CC(=O)NCCCC(=O)O

Specifications

Melting Point 129°C to 131°C
Beilstein 1762288
Solubility Information Soluble in water (19.9mg/ml).
Quantity 250 g
Formula Weight 145.16
Percent Purity 99%
Chemical Name or Material 4-Acetamidobutyric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-186-1
RTECSNumber ES5610000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.