Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. A1025250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1025250G 250 g
A102525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1025250G Supplier TCI America Supplier No. A1025250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92-91-1
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00008749
InChI Key QCZZSANNLWPGEA-UHFFFAOYSA-N
Synonym 4-acetylbiphenyl, 4'-phenylacetophenone, 4-phenylacetophenone, 4-biphenylyl methyl ketone, p-phenylacetophenone, p-acetylbiphenyl, 1-1,1'-biphenyl-4-yl ethanone, acetophenone, 4'-phenyl, 1-4-phenylphenyl ethanone, ketone, 4-biphenylyl methyl
PubChem CID 7113
IUPAC Name 1-(4-phenylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 122°C
Color White-Yellow
Boiling Point 168°C
Quantity 250 g
Formula Weight 196.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetylbiphenyl
RTECSNumber AM9662502
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.