Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylpyridazine, 95%, Thermo Scientific™

Catalog No. AAH6646103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6646103 1 g
AAH66461MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6646103 Supplier Thermo Scientific Chemicals Supplier No. H6646103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50901-46-7
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD13193354
InChI Key IWFYWGOLMZIBQB-UHFFFAOYSA-N
Synonym 1-pyridazin-4-yl ethanone, 1-pyridazin-4-yl ethan-1-one, 4-acetylpyridazine, ethanone, 1-4-pyridazinyl, 4-acetyl-pyridazin, 1-pyridazin-4-yl-ethanone, methyl 4-pyridazinyl ketone, 1-pyridazin-4-yl ;ethanone, 1-4-pyridazinyl-ethanone, ethanone,1-4-pyridazinyl-9ci
PubChem CID 14452005
IUPAC Name 1-pyridazin-4-ylethanone
SMILES CC(=O)C1=CN=NC=C1

Specifications

Quantity 1 g
Beilstein 742138
Formula Weight 122.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Acetylpyridazine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.