Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acryloylmorpholine (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. A0841100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0841100ML 100 mL
A084125ML 25 mL
A0841500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0841100ML Supplier TCI America Supplier No. A0841100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5117-12-4
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00047413
InChI Key XLPJNCYCZORXHG-UHFFFAOYSA-N
Synonym 4-acryloylmorpholine, 1-morpholinoprop-2-en-1-one, n-acrylylmorpholine, n-acryloylmorpholine, morpholine, 4-1-oxo-2-propenyl, poly acryloylmorpholine, acryloylmorpholine, acryloyl morpholine, 1-morpholin-4-yl prop-2-en-1-one, unii-k0y58p61ja
PubChem CID 98723
IUPAC Name 1-morpholin-4-ylprop-2-en-1-one
SMILES C=CC(=O)N1CCOCC1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 100 mL
Formula Weight 141.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Acryloylmorpholine (stabilized with MEHQ)
EINECSNumber (5)-0865
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.