Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(alpha-Methylbenzyl)resorcinol 98.0+%, TCI America™
SDP

Catalog No. M27541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27541G 1 g
M27545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27541G Supplier TCI America Supplier No. M27541G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-27-8
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD21648466
InChI Key PQSXNIMHIHYFEE-UHFFFAOYNA-N
Synonym 4-(1-Phenylethyl)benzene-1,3-diol, 4-(1-Phenylethyl)resorcinol
PubChem CID 11171903
IUPAC Name 4-(1-phenylethyl)benzene-1,3-diol
SMILES CC(C1=CC=CC=C1)C1=C(O)C=C(O)C=C1

Specifications

Melting Point 81°C
Boiling Point 244°C
Quantity 1 g
Formula Weight 214.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(alpha-Methylbenzyl)resorcinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.