Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-benzylpiperidine 98.0+%, TCI America™
SDP

Catalog No. A1086100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1086100G 100 g
A108625G 25 g
A1086500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1086100G Supplier TCI America Supplier No. A1086100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50541-93-0
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00006504
InChI Key YUBDLZGUSSWQSS-UHFFFAOYSA-N
Synonym 4-amino-1-benzylpiperidine, 1-benzyl-4-aminopiperidine, 1-benzyl-piperidin-4-ylamine, n-benzyl-4-amino piperidine, 1-benzyl-4-piperidylamine, 4-piperidinamine, 1-phenylmethyl, 4-amino-1-n-benzylpiperidine, 4-amino-n-benzylpiperidine, 1-phenylmethyl-4-piperidinamine, n-benzyl-4-aminopiperidine
PubChem CID 415852
IUPAC Name 1-benzylpiperidin-4-amine
SMILES C1CN(CCC1N)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 100 g
Formula Weight 190.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Amino-1-benzylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.