Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-butanol, 98%, Thermo Scientific™

Catalog No. AA4152603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4152603 1 g
AA4152606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AA4152603 Supplier Thermo Scientific Chemicals Supplier No. 4152603
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13325-10-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00008230
InChI Key BLFRQYKZFKYQLO-UHFFFAOYSA-N
Synonym 4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
PubChem CID 25868
IUPAC Name 4-aminobutan-1-ol
SMILES C(CCO)CN

Specifications

Density 0.967g/mL
Boiling Point 206°C
Flash Point 107° (226°F)
Refractive Index 1.4610
Quantity 1 g
UN Number UN2735
Formula Weight 89.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Amino-1-butanol
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.