Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1H-imidazole-5-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. A128210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A128210G 10 g
A128225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A128210G Supplier TCI America Supplier No. A128210G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5098-11-3
Molecular Formula C4H4N4
Molecular Weight (g/mol) 108.10
MDL Number MFCD00129663
InChI Key XEPBRDBFOSKYCF-UHFFFAOYSA-N
Synonym 4(5)-Amino-5(4)-cyanoimidazole
PubChem CID 78777
IUPAC Name 4-amino-1H-imidazole-5-carbonitrile
SMILES C1=NC(=C(N1)C#N)N

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 10 g
UN Number 3439
Formula Weight 108.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-1H-imidazole-5-carbonitrile
EINECSNumber (5)-0389
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.