Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2,6-dimethylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. A209125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A209125G 25 g
A20915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A209125G Supplier TCI America Supplier No. A209125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 461-98-3
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.159
MDL Number MFCD00006106
InChI Key BJJDXAFKCKSLTE-UHFFFAOYSA-N
Synonym 4-amino-2,6-dimethylpyrimidine, kyanmethin, cyanomethine, cyanmethine, kyanmethine, 2,6-dimethyl-4-pyrimidinamine, 2,4-dimethyl-6-aminopyrimidine, 6-amino-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinylamine, 4-pyrimidinamine, 2,6-dimethyl
PubChem CID 68039
IUPAC Name 2,6-dimethylpyrimidin-4-amine
SMILES CC1=CC(=NC(=N1)C)N

Specifications

Melting Point 185°C
Color White-Yellow
Boiling Point 254°C
Quantity 25 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,6-dimethylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.