Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-chlorobenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. A095125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A095125G 25 g
A09515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A095125G Supplier TCI America Supplier No. A095125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2457-76-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007772
InChI Key MBDUKNCPOPMRJQ-UHFFFAOYSA-N
Synonym 2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109
PubChem CID 17154
ChEBI CHEBI:59472
IUPAC Name 4-amino-2-chlorobenzoic acid
SMILES C1=CC(=C(C=C1N)Cl)C(=O)O

Specifications

Melting Point 211°C
Quantity 25 g
Formula Weight 171.58
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chlorobenzoic Acid
EINECSNumber (3)-1480
RTECSNumber DG1575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.