Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-chlorophenol, 99%

Catalog No. AAB2342006
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2342006 5 g
AAB2342003 1 g
AAB2342014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2342006 Supplier Thermo Scientific Chemicals Supplier No. B2342006
Add to Cart

CAS: 3964-52-1 | C6H6ClNO | 143.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3964-52-1
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD00128891
InChI Key ZYZQSCWSPFLAFM-UHFFFAOYSA-N
Synonym 3-chloro-4-hydroxyaniline, 2-chloro-4-aminophenol, phenol, 4-amino-2-chloro, 4-amino-2-chloro-phenol, unii-2z6zt3b98u, o-chloro-p-aminophenol, pubchem18907, salzsaurem p-aminophenol, acmc-1aiqv, 3-chloro4-hydroxyaniline
PubChem CID 77578
IUPAC Name 4-amino-2-chlorophenol
SMILES NC1=CC=C(O)C(Cl)=C1

Specifications

Melting Point 150°C to 153°C
Quantity 5 g
Formula Weight 143.57
Percent Purity 99%
Chemical Name or Material 4-Amino-2-chlorophenol
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.