Learn More
Sigma Organic Chemistry 4-Amino-2-methoxyphenol | 1G | 52200-90-5 | MFCD01707809
Supplier: Sigma Organic Chemistry CBR016181G

4-Amino-2-methoxyphenol , 1G
About this Item:
CAS #: 52200-90-5
MDL #: MFCD01707809
Molecular Weight: 139.15
UNSPSC Code: 12352200
Chemical Formula: C7H9NO2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.