Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-nitrophenol 95.0+%, TCI America™
SDP

Catalog No. A037925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A037925G 25 g
A03795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A037925G Supplier TCI America Supplier No. A037925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 119-34-6
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00007876
InChI Key WHODQVWERNSQEO-UHFFFAOYSA-N
Synonym phenol, 4-amino-2-nitro, fourrine 57, 4-hydroxy-3-nitroaniline, 2-nitro-4-aminophenol, fourrine brown pr, oxidation base 25, fourrine brown propyl, o-nitro-p-aminophenol, p-amino-o-nitrophenol, c.i. oxidation base 25
PubChem CID 3417419
ChEBI CHEBI:82385
IUPAC Name 4-amino-2-nitrophenol
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])O

Specifications

Melting Point 129°C
Quantity 25 g
Formula Weight 154.13
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-nitrophenol
EINECSNumber (3)-0788
RTECSNumber SJ6303000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.