Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%

Catalog No. AAH5234903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5234903 1 g
AAH52349MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5234903 Supplier Thermo Scientific Chemicals Supplier No. H5234903
Add to Cart

CAS: 819058-34-9 | C12H17BFNO2 | 237.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 819058-34-9
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.08
MDL Number MFCD09033884
InChI Key AIXGNRNTXUKZLC-UHFFFAOYSA-N
Synonym 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid, pinacol ester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronicacidpinacolester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 17906172
IUPAC Name 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 237.08
Percent Purity 96%
Chemical Name or Material 4-Amino-3-fluorobenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.