Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-hydroxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A119425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A119425G 25 g
A11945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A119425G Supplier TCI America Supplier No. A119425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2374-03-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00017094
InChI Key NFPYJDZQOKCYIE-UHFFFAOYSA-N
Synonym 3-hydroxy-4-aminobenzoic acid, 4-amino-3-hydroxy benzoic acid, benzoic acid, 4-amino-3-hydroxy, chembl1462, 2hdr, pubchem14418, 2-amino-5-carboxyphenol, intermediates-zcf02635, acmc-209g5k, 4-carboxy-2-hydroxyaniline
PubChem CID 137566
IUPAC Name 4-amino-3-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)O)N

Specifications

Quantity 25 g
Formula Weight 153.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-hydroxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.