Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. A226025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A226025G 25 g
A22605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A226025G Supplier TCI America Supplier No. A226025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1681-37-4
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.114
MDL Number MFCD00160304
InChI Key IUPPEELMBOPLDJ-UHFFFAOYSA-N
Synonym 4-amino-3-nitropyridine, 4-pyridinamine, 3-nitro, 3-nitro-4-pyridinamine, 3-nitro-4-aminopyridine, 3-nitro-pyridin-4-ylamine, 3-nitro-4-pyridylamine, pubchem1277, acmc-209dxo, 3-nitro-4-amino pyridine, 3-nitro-4-pyridinamine #
PubChem CID 548803
IUPAC Name 3-nitropyridin-4-amine
SMILES C1=CN=CC(=C1N)[N+](=O)[O-]

Specifications

Melting Point 205°C
Color Yellow
Quantity 25 g
Formula Weight 139.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.