Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-bromo-2-hydroxypyrimidine, 98%

Catalog No. AAH6195203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6195203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6195203 Supplier Thermo Scientific Chemicals Supplier No. H6195203

missing translation for 'validMainframeMsg'

CAS: 2240-25-7 | C4H4BrN3O | 190.00 g/mol

4-Amino-5-bromo-2-hydroxypyrimidine can be used in the synthesis of cross-link products under anaerobic and aerobic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-5-bromo-2-hydroxypyrimidine can be used in the synthesis of cross-link products under anaerobic and aerobic conditions.

Solubility
Soluble in formic acid (50 mg/ml).

Notes
Store away from st
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2240-25-7
Molecular Formula C4H4BrN3O
Molecular Weight (g/mol) 190.00
MDL Number MFCD00056025
InChI Key QFVKLKDEXOWFSL-UHFFFAOYSA-N
Synonym 5-bromocytosine, 4-amino-5-bromopyrimidin-2 1h-one, 6-amino-5-bromopyrimidin-2 1h-one, 4-amino-5-bromo-2-pyrimidinol, 4-amino-5-bromopyrimidin-2-ol, 2 1h-pyrimidinone, 4-amino-5-bromo, 4-amino-5-bromo-2-hydroxypyrimidine, 2 1h-pyrimidinone,6-amino-5-bromo, 6-amino-5-bromo-1,2-dihydropyrimidin-2-one, 4-amino-5-bromo-1,2-dihydropyrimidin-2-one
PubChem CID 75233
IUPAC Name 6-amino-5-bromo-1H-pyrimidin-2-one
SMILES NC1=C(Br)C=NC(=O)N1

Specifications

Melting Point 240°C to 243°C
Color White to Yellow
Quantity 1 g
Beilstein 127286
Solubility Information Soluble in formic acid (50mg/ml).
Formula Weight 190
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Amino-5-bromo-2-hydroxypyrimidine
Hazard Category H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
EINECSNumber 218-806-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.