Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-bromo-6-chloropyrimidine, 98%, Thermo Scientific™

Catalog No. AAH64369MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64369MD 250 mg
AAH6436903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64369MD Supplier Thermo Scientific Chemicals Supplier No. H64369MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 663193-80-4
Molecular Formula C4H3BrClN3
Molecular Weight (g/mol) 208.443
MDL Number MFCD09999224
InChI Key DQSBRYJETNYGCI-UHFFFAOYSA-N
Synonym 4-amino-5-bromo-6-chloropyrimidine, 5-bromo-6-chloro-4-pyrimidinamine, 5-bromo-6-chloro-pyrimidin-4-ylamine
PubChem CID 45789713
IUPAC Name 5-bromo-6-chloropyrimidin-4-amine
SMILES C1=NC(=C(C(=N1)Cl)Br)N

Specifications

Color Gray
Quantity 250 mg
Beilstein 23483365
Formula Weight 208.44
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Amino-5-bromo-6-chloropyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.