Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-chloro-2,6-dimethylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. A19015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19015G 5 g
A190125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19015G Supplier TCI America Supplier No. A19015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2858-20-0
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.60
MDL Number MFCD00191727
InChI Key BKFCZKYCVQQMCX-UHFFFAOYSA-N
Synonym 4-amino-5-chloro-2,6-dimethylpyrimidine, timtec-bb sbb004000, 4-amino-2-chloro-6,7-dimethylpyrimidine, 5-chloro-2,6-dimethylpyrimidine-4-ylamine, acmc-209h3e, 5-chloro-2,6-dimethyl-4-pyrimidinamine, 2,4-dimethyl-5-chloro-6-aminopyrimidine, 2,6-dimethyl-4-amino-5-chloropyrimidine, 4-pyrimidinamine,5-chloro-2,6-dimethyl, 5-chloro-2,6-dimethyl-4-pyrimidinylamine
PubChem CID 593843
IUPAC Name 5-chloro-2,6-dimethylpyrimidin-4-amine
SMILES CC1=NC(C)=C(Cl)C(N)=N1

Specifications

Melting Point 165°C
Quantity 5 g
Formula Weight 157.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-5-chloro-2,6-dimethylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.