Learn More
4-Amino-5-cyano-2-methylpyrimidine, 95%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
4-Amino-5-cyano-2-methylpyrimidine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water (6.4 g/L at 25°C). Soluble in dimethyl sulfoxide, ethanol and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| UN Number | UN3439 |
| Solubility Information | Slightly soluble in water (6.4g/L at 25°C). Soluble in dimethyl sulfoxide,ethanol and methanol. |
| Formula Weight | 134.14 |
| Percent Purity | 95% |
| Physical Form | Powder |
| Chemical Name or Material | 4-Amino-5-cyano-2-methylpyrimidine |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Hazard Statement | GHS H Statement H302-H312-H332-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S. |
| EINECSNumber | 211-814-2 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.