Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-6-chloropyrimidine 98.0+%, TCI America™
SDP

Catalog No. A21361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21361G 1 g
A21365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21361G Supplier TCI America Supplier No. A21361G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5305-59-9
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD00053576
InChI Key DUKKRSPKJMHASP-UHFFFAOYSA-N
Synonym 4-amino-6-chloropyrimidine, 6-chloropyrimidin-4-ylamine, 6-chloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 6-chloro, 6-chloro-4-pyrimidinamine, 6-amino-4-chloropyrimidine, 6-chloropyrimidine-4-ylamine, 6-chloro-4-aminopyrimidine, 6-chlorpyrimidin-4-amin, zlchem 630
PubChem CID 238012
IUPAC Name 6-chloropyrimidin-4-amine
SMILES C1=C(N=CN=C1Cl)N

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 1 g
Formula Weight 129.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-chloropyrimidine
EINECSNumber (9)-2241
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.