Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-N-methylphthalimide 98.0+%, TCI America™
SDP

Catalog No. A156125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A156125G 25 g
A15615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A156125G Supplier TCI America Supplier No. A156125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2307-00-8
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD00012081
InChI Key KMEBUNSLFRQSEM-UHFFFAOYSA-N
PubChem CID 726931
IUPAC Name 5-amino-2-methylisoindole-1,3-dione
SMILES CN1C(=O)C2=C(C1=O)C=C(C=C2)N

Specifications

Melting Point 247°C
Quantity 25 g
Formula Weight 176.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-N-methylphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.