Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-N-(tert-butoxycarbonyl)-D-phenylalanine 97.0+%, TCI America™
SDP

Catalog No. A21301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21301G 1 g
A21305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21301G Supplier TCI America Supplier No. A21301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 164332-89-2
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.324
MDL Number MFCD00236818
InChI Key NDMVQEZKACRLDP-LLVKDONJSA-N
Synonym boc-4-amino-d-phenylalanine, boc-d-phe 4-nh2-oh, boc-p-amino-d-phe-oh, boc-4-amino-d-phe-oh, boc-d-4-aminophenylalanine, boc-d-4-aminophe, r-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 4-amino-n-tert-butoxycarbonyl-d-phenylalanine, 2r-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, d-phenylalanine, 4-amino-n-1,1-dimethylethoxy carbonyl
PubChem CID 7019658
IUPAC Name (2R)-3-(4-aminophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)N)C(=O)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 280.32
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-N-(tert-butoxycarbonyl)-D-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.