Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzoic Acid 99.0+%, TCI America™
SDP

Catalog No. A0269100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0269100G 100 g
A026925G 25 g
A0269500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0269100G Supplier TCI America Supplier No. A0269100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 150-13-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007894
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
PubChem CID 978
ChEBI CHEBI:30753
IUPAC Name 4-aminobenzoic acid
SMILES NC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 189°C
pH 3.5 (0.5% soln.)
Quantity 100 g
Formula Weight 137.14
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobenzoic Acid
EINECSNumber (3)-1454
RTECSNumber DG1400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.