Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzonitrile, 98%

Catalog No. AAA1379409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1379409 10 g
AAA1379418 50 g
AAA1379430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1379409 Supplier Thermo Scientific Chemicals Supplier No. A1379409
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 873-74-5 | C7H6N2 | 118.139 g/mol

4-Aminobenzonitrile is used as an amino substituted benzonitrile with hypotensive activity and as a radioprotective agent. It is also used as a derivatization reagent in the capillary zone electrophoretic analysis of aldoses, ketoses and uronic acid. Further, it is used in the preparation of methacrylic monomers containing pendant azobenzene structures and polythiophenes containing an azobenzene moiety in the side-chain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzonitrile is used as an amino substituted benzonitrile with hypotensive activity and as a radioprotective agent. It is also used as a derivatization reagent in the capillary zone electrophoretic analysis of aldoses, ketoses and uronic acid. Further, it is used in the preparation of methacrylic monomers containing pendant azobenzene structures and polythiophenes containing an azobenzene moiety in the side-chain.

Solubility
Soluble in ethyl acetate, dichloromethane and chloroform. Insoluble in water.

Notes
Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-74-5
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00007821
InChI Key YBAZINRZQSAIAY-UHFFFAOYSA-N
Synonym 4-cyanoaniline, p-aminobenzonitrile, p-cyanoaniline, benzonitrile, 4-amino, aniline, p-cyano, benzonitrile, p-amino, 1-amino-4-cyanobenzene, unii-pb8v69y76t, 4-amino benzonitrile, 4-amino-benzonitrile
PubChem CID 13396
IUPAC Name 4-aminobenzonitrile
SMILES C1=CC(=CC=C1C#N)N

Specifications

Melting Point 83°C to 87°C
Quantity 10 g
UN Number UN3439
Beilstein 774507
Solubility Information Soluble in ethyl acetate,dichloromethane and chloroform. Insoluble in water.
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 4-Aminobenzonitrile
Hazard Category H301-H315-H319
Hazard Statement GHS H Statement
H301-H312-H319
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 212-850-1
RTECSNumber BX2320000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.