Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobiphenyl Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USNH1301
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-NH-1301 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-NH-1301 Supplier Agilent Technologies Supplier No. NH1301
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92-67-1 , 67-56-1
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
InChI Key DMVOXQPQNTYEKQ-UHFFFAOYSA-N
PubChem CID 7102
ChEBI CHEBI:1784
IUPAC Name 4-phenylaniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N

Specifications

CAS Max % 99.99, 0.01
Analyte or Component Names Methanol,4-aminobiphenyl
Element Methanol, 4-aminobiphenyl
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material 4-Aminobiphenyl Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.