Learn More
Selleck Chemical LLC 4-Aminobutyric acid GABA-100mg
SDP
Supplier:  Selleck Chemical LLC S4700100MG
Encompass_Preferred
4-Aminobutyric acid (4-Aminobutanoic acid, GABA, Gamma-aminobutyric acid, Piperidic acid) is a naturally occurring neurotransmitter with central nervous system (CNS) inhibitory activity.
Catalog No. 50-238-9725
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.