Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Aminohexanophenone 98.0+%, TCI America™
SDP

Catalog No. A205825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A205825G 25 g
A20585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A205825G Supplier TCI America Supplier No. A205825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38237-76-2
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
InChI Key YDQJFPLRVSPPGE-UHFFFAOYSA-N
Synonym 4-Hexanoylaniline
PubChem CID 37983
IUPAC Name 1-(4-aminophenyl)hexan-1-one
SMILES CCCCCC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 85°C
Quantity 25 g
UN Number 2811
Formula Weight 191.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Aminohexanophenone
RTECSNumber MP4450000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.