Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoisoquinoline 98.0+%, TCI America™
SDP

Catalog No. A25951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25951G 1 g
A25955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25951G Supplier TCI America Supplier No. A25951G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23687-25-4
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00034752
InChI Key ISIUXVGHQFJYHM-UHFFFAOYSA-N
PubChem CID 90237
IUPAC Name isoquinolin-4-amine
SMILES C1=CC=C2C(=C1)C=NC=C2N

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminoisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.