Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminomethyltetrahydropyran, 97%, Acros Organics

Click to view available options
Quantity:
250mg
1g
5g
25g
130290-79-8
C6H13NO
115.17
MFCD02179435
IPBPLHNLRKRLPJ-UHFFFAOYSA-N
4-aminomethyltetrahydropyran,4-aminomethyl tetrahydro-2h-pyran,tetrahydro-2h-pyran-4-yl methanamine,4-aminomethyl tetrahydropyran,1-tetrahydro-2h-pyran-4-ylmethanamine,tetrahydropyran-4-yl methylamine,c-tetrahydro-pyran-4-yl-methylamine,tetrahydro-2h-pyran-4-yl methylamine,2h-pyran-4-methanamine, tetrahydro,oxan-4-yl methanamine
2773210
oxan-4-ylmethanamine
C1COCCC1CN
130290-79-8
C6H13NO
115.17
MFCD02179435
IPBPLHNLRKRLPJ-UHFFFAOYSA-N
4-aminomethyltetrahydropyran,4-aminomethyl tetrahydro-2h-pyran,tetrahydro-2h-pyran-4-yl methanamine,4-aminomethyl tetrahydropyran,1-tetrahydro-2h-pyran-4-ylmethanamine,tetrahydropyran-4-yl methylamine,c-tetrahydro-pyran-4-yl-methylamine,tetrahydro-2h-pyran-4-yl methylamine,2h-pyran-4-methanamine, tetrahydro,oxan-4-yl methanamine
2773210
oxan-4-ylmethanamine
C1COCCC1CN
Specific Gravity 1.02
Quantity 25g
Boiling Point 75.0°C to 76.0°C (18.0mmHg)
Density 1.0200g/mL
Refractive Index 1.4640 to 1.4660
Assay Percent Range 96% min. (GC)
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material 4-Aminomethyltetrahydropyran

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger