Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminononafluorobiphenyl 96.0+%, TCI America™
SDP

Catalog No. A13461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13461G 1 g
A13465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13461G Supplier TCI America Supplier No. A13461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 969-25-5
Molecular Formula C12H2F9N
Molecular Weight (g/mol) 331.141
MDL Number MFCD00671483
InChI Key DVKUPHGYOMHVDR-UHFFFAOYSA-N
PubChem CID 630342
IUPAC Name 2,3,5,6-tetrafluoro-4-(2,3,4,5,6-pentafluorophenyl)aniline
SMILES C1(=C(C(=C(C(=C1F)F)N)F)F)C2=C(C(=C(C(=C2F)F)F)F)F

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 1 g
Formula Weight 331.14
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminononafluorobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.