Learn More
Sigma Organic Chemistry 4-aminophenyl methyl su | 50mg | MFCD00025353
Supplier: Sigma Organic Chemistry CDS01266150MG

4-aminophenyl methyl su , 50mg
About this Item:
MDL #: MFCD00025353
Molecular Weight: 171.22
UNSPSC Code: 12352200
Chemical Formula: C7H9NO2S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.