Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 4-Aminophenyl Phosphate Monosodium Salt, >99% Purity, Solid, 10 mg

Catalog No. 502706404
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6404 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6404 Supplier Abcam Supplier No. AB14189910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Highly pure substrate for electrochemical alkaline phosphatase measurement.

  • Molecular weight: 211.09 Da
  • Purity: >99%
  • Physical state: Solid
  • Application: Substrate for electrochemical alkaline phosphatase measurement
  • Storage buffer: 100% sodium (4-aminophenyl) hydrogen phosphate
  • Storage temperature: -20°C
  • Shipping temperature: Ambient (can ship with ice)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Alkaline Phosphatase
Substrate Properties Chemical Substrate
Substrate 4-Aminophenyl phosphate
Shipping Condition Ambient
Reactivity Alkaline Phosphatase
Content And Storage Store at -20°C.
Buffer 100% sodium (4-aminophenyl) hydrogen phosphate
Form Solid
Formulation 100% sodium (4-aminophenyl) hydrogen phosphate
Quantity 10 mg
For Use With (Application) Electrochemical alkaline phosphatase measurement
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.