Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 4-Aminophenyl Phosphate Monosodium Salt, Alkaline Phosphatase Substrate, 50 mg

Catalog No. 502706403
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6403 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6403 Supplier Abcam Supplier No. AB14189950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity substrate for electrochemical alkaline phosphatase measurement.

  • Purity: >99%
  • Molecular Weight: 211.09 Da
  • Form: Solid
  • Application: Substrate for electrochemical alkaline phosphatase measurement

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Alkaline Phosphatase
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Reactivity Alkaline Phosphatase
Content And Storage Store at -20°C.
Form Solid
Buffer 100% sodium (4-aminophenyl) hydrogen phosphate
Formulation 100% sodium (4-aminophenyl) hydrogen phosphate
Quantity 50 mg
For Use With (Application) Electrochemical alkaline phosphatase measurement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.