Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminophenylacetic acid, 98%

Catalog No. AC104280250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104280250 25 g Glass Bottle
AC104280050 5 g Glass Bottle
AC104281000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104280250 Supplier Thermo Scientific Chemicals Supplier No. 104280250
Add to Cart

CAS: 1197-55-3 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197-55-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007916
InChI Key CSEWAUGPAQPMDC-UHFFFAOYSA-N
Synonym 4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
PubChem CID 14533
IUPAC Name 2-(4-aminophenyl)acetic acid
SMILES NC1=CC=C(CC(O)=O)C=C1

Specifications

Melting Point 196.0°C to 201.0°C
CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula H2NC6H4CH2CO2H
Beilstein 14, 456
Merck Index 15, 459
Water 1% max. (K.F.)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies, soluble in alcohol
Quantity 25 g
Formula Weight 151.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Aminophenylacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 214-828-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.