Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminopyridine, ∽98%, MP Biomedicals™

Catalog No. ICN15036680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15036680 100 g
ICN15036605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15036680 Supplier MP Biomedicals, Inc Supplier No. 0215036680
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 504-24-5 | C5H6N2 | 94.117 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-24-5
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
InChI Key NUKYPUAOHBNCPY-UHFFFAOYSA-N
Synonym 4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
PubChem CID 1727
ChEBI CHEBI:34385
IUPAC Name pyridin-4-amine
SMILES C1=CN=CC=C1N

Specifications

Color Yellow
Assay Percent Range ∽98%
Quantity 100 g
Formula Weight 94.13
Percent Purity 98%
Grade Not Available
Physical Form Crystals
Chemical Name or Material 4-Aminopyridine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.