CAS RN 504-24-5
4-Aminopyridine, ∽98%, MP Biomedicals™
CAS: 504-24-5 Molecular Formula: C5H6N2 Molecular Weight (g/mol): 94.117 InChI Key: NUKYPUAOHBNCPY-UHFFFAOYSA-N Synonym: 4-aminopyridine,fampridine,4-pyridinamine,dalfampridine,4-pyridylamine,avitrol,p-aminopyridine,4-ap,ampyra,gamma-aminopyridine PubChem CID: 1727 ChEBI: CHEBI:34385 IUPAC Name: pyridin-4-amine SMILES: C1=CN=CC=C1N
4-Aminopyridine, 99+%
CAS: 504-24-5 Molecular Formula: C5H6N2 Molecular Weight (g/mol): 94.117 MDL Number: MFCD00006439 InChI Key: NUKYPUAOHBNCPY-UHFFFAOYSA-N Synonym: 4-aminopyridine,fampridine,4-pyridinamine,dalfampridine,4-pyridylamine,avitrol,p-aminopyridine,4-ap,ampyra,gamma-aminopyridine PubChem CID: 1727 ChEBI: CHEBI:34385 IUPAC Name: pyridin-4-amine SMILES: C1=CN=CC=C1N
4-Aminopyridine, 98%
CAS: 504-24-5 Molecular Formula: C5H6N2 Molecular Weight (g/mol): 94.12 MDL Number: MFCD00006439 InChI Key: NUKYPUAOHBNCPY-UHFFFAOYSA-N Synonym: 4-aminopyridine,fampridine,4-pyridinamine,dalfampridine,4-pyridylamine,avitrol,p-aminopyridine,4-ap,ampyra,gamma-aminopyridine PubChem CID: 1727 ChEBI: CHEBI:34385 IUPAC Name: pyridin-4-amine SMILES: C1=CN=CC=C1N
4-Aminopyridine, 98%
CAS: 504-24-5 Molecular Formula: C5H6N2 Molecular Weight (g/mol): 94.117 MDL Number: MFCD00006439 InChI Key: NUKYPUAOHBNCPY-UHFFFAOYSA-N Synonym: 4-aminopyridine,fampridine,4-pyridinamine,dalfampridine,4-pyridylamine,avitrol,p-aminopyridine,4-ap,ampyra,gamma-aminopyridine PubChem CID: 1727 ChEBI: CHEBI:34385 IUPAC Name: pyridin-4-amine SMILES: C1=CN=CC=C1N
4-Aminopyridine 99.0+%, TCI America™
CAS: 504-24-5 Molecular Formula: C5H6N2 Molecular Weight (g/mol): 94.117 MDL Number: MFCD00006439 InChI Key: NUKYPUAOHBNCPY-UHFFFAOYSA-N Synonym: 4-aminopyridine,fampridine,4-pyridinamine,dalfampridine,4-pyridylamine,avitrol,p-aminopyridine,4-ap,ampyra,gamma-aminopyridine PubChem CID: 1727 ChEBI: CHEBI:34385 IUPAC Name: pyridin-4-amine SMILES: C1=CN=CC=C1N
R&D Systems™ 4-Aminopyridine
CAS: 504-24-5 Molecular Formula: C5H6N2
Sigma Aldrich 4-Aminopyridine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 4-Aminopyridine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.