Learn More
Sigma Organic Chemistry 4-aminosulfonyl benzene | 100mg | MFCD07352256
Supplier: Sigma Organic Chemistry CDS011892100MG

4-aminosulfonyl benzene , 100mg
About this Item:
MDL #: MFCD07352256
Molecular Weight: 255.7
UNSPSC Code: 12352200
Chemical Formula: C6H6ClNO4S2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.