Learn More
Medchemexpress LLC 4-Arm PEG-SH MW 500 100mg
SDP
Supplier:  Medchemexpress LLC HYW1048849A100MG
Encompass_Preferred
4-Arm PEG-SH (MW 5000) (4-Arm PEG-thiol (MW 5000)) is a multi-arm PEG derivative with thiol groups at each end of the four arms The reactive free thiol SH or sulfhydryl groups react selectively with maleimide and transition metal surfaces including gold and silver[1]
Catalog No. 50-002-05020