Learn More
Medchemexpress LLC 4-Arm PEG-SH MW 500 25mg
SDP
Supplier:  Medchemexpress LLC 25MGHYW1048849A
Encompass_Preferred
4-Arm PEG-SH (MW 5000) (4-Arm PEG-thiol (MW 5000)) is a multi-arm PEG derivative with thiol groups at each end of the four arms The reactive free thiol SH or sulfhydryl groups react selectively with maleimide and transition metal surfaces including gold and silver[1]
Catalog No. 50-002-05856