Learn More
AdipoGen 4-Azido-7-nitrobenzofurazan
SDP
Supplier:  AdipoGen CDXA0177M100
Encompass_Preferred
Chemical. CAS 10199-90-3. Formula C6H2N6O3. MW 206.12. Synthetic. Non-fluorescent azido fluorogen which can be photoactivated to the longer wavelength amino version that is fluorescent. Building block and reagent for synthesis of fluorescent probes. Used for the preparation of fluorogenic substrates, chemosensors and probes, which can be concerted to the high fluoresence NBD-amine after release. Fluorescent chemosensor for hydrogen sulfide. Spectral data Ex=384 454nm, Em=550nm.
Catalog No. 50-256-1816
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.