Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzamidopiperidine Hydrate 98.0+%, TCI America™
SDP

Catalog No. B302525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B302525G 25 g
B30255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B302525G Supplier TCI America Supplier No. B302525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33953-37-6
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.27
MDL Number MFCD01314226
InChI Key JMQDNLCNCDSHNC-UHFFFAOYSA-N
Synonym n-piperidin-4-yl-benzamide, n-piperidin-4-yl benzamide, n-4-piperidyl benzamide, 4-benzamidopiperidine, n-4-piperidylbenzamide, benzamide, n-4-piperidinyl, 4-benzimido piperidine, n-4-piperidinyl benzamide, 4-ben-zamidopiperidine
PubChem CID 118120
IUPAC Name N-(piperidin-4-yl)benzamide
SMILES O=C(NC1CCNCC1)C1=CC=CC=C1

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 25 g
Formula Weight 204.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzamidopiperidine Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.