Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzoylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B116425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B116425G 25 g
B11645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B116425G Supplier TCI America Supplier No. B116425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 611-95-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00002560
InChI Key IFQUPKAISSPFTE-UHFFFAOYSA-N
Synonym p-benzoylbenzoic acid, benzoic acid, 4-benzoyl, 4-carboxybenzophenone, 4-benzoyl-benzoic acid, 4-phenylcarbonyl benzoic acid, benzoic acid, p-benzoyl, benzophenone-4-carboxylic acid, 4-benzoyl benzoic acid, pubchem14544, acmc-209mpx
PubChem CID 69147
IUPAC Name 4-benzoylbenzoic acid
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 196°C
Color White
Quantity 25 g
Formula Weight 226.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzoylbenzoic Acid
RTECSNumber DG3650000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.