Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Benzyl-1,4-oxazinan-2-yl)methanol, Thermo Scientific™

Catalog No. CC12409CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC12409CB 250 mg
CC12409DA 1 g
CC12409DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC12409CB Supplier Thermo Scientific Chemicals Supplier No. CC12409CB
Add to Cart

Chemical Identifiers

CAS 40987-24-4
Molecular Formula C12H18NO2
Molecular Weight (g/mol) 208.28
MDL Number MFCD02682009,MFCD10698663,MFCD11848685
InChI Key WQNIKIMRIXHNFF-GFCCVEGCSA-O
Synonym 4-benzylmorpholin-2-yl methanol, 4-benzyl-1,4-oxazinan-2-yl methanol, 4-benzyl-2-hydroxymethyl morpholine, 2-hydroxymethyl-4-benzylmorpholine, 2-morpholinemethanol, 4-phenylmethyl, 4-benzyl-morpholin-2-yl-methanol, 4-phenylmethyl-2-morpholinyl methanol, 4-phenylmethyl morpholin-2-yl methanol, 2-morpholinemethanol,4-phenylmethyl-, 2s, acmc-20a6qw
PubChem CID 2776358
SMILES OC[C@H]1C[NH+](CC2=CC=CC=C2)CCO1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name (4-benzylmorpholin-2-yl)methanol
Formula Weight 207.27
Percent Purity 95%
Chemical Name or Material (4-Benzyl-1, 4-oxazinan-2-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.